Orders LinkContactUsLinkComponent
1,4-Dithioerythritol
1,4-Dithioerythritol
Glass bottle for solids, 1 g
  SERA39758.01
Marketsource
 :   Serva
1,4-Dithioerythritol
  SERA39758.01
 :  Serva
Marketsource
 :  6892-68-8
1,4-Dithioerythritol
Glass bottle for solids, 1 g
 :  6892-68-8
1 * 1 g
 :  Cleland's Reagent, (R*,S*)-1,4-Dimercaptobutane-2,3-diol, DTE, Erythro-1,4-Dimercaptobutane-2,3-diol, trans-1,4-Dimercaptobutane-2,3-diol, Dithioerythritol, DL-Dithioerythritol
  •  :6892-68-8
  •  :C₄H₁₀O₂S₂
  •  :MFCD00063750
  •  :154.25 g/mol
  •  :286 °C (1013 hPa)
  •  :82…83 °C
  •  :Kamertemperatuur
  •  :229-998-8
  • Verpakking:
    Glass bottle for solids
  • VPE:
    1 g