1,4-Dithioerythritol
Glass bottle for solids, 1 g
SERA39758.01
Marketsource
: Serva
: Cleland's Reagent, (R*,S*)-1,4-Dimercaptobutane-2,3-diol, DTE, Erythro-1,4-Dimercaptobutane-2,3-diol, trans-1,4-Dimercaptobutane-2,3-diol, Dithioerythritol, DL-Dithioerythritol
- :6892-68-8
- :C₄H₁₀O₂S₂
- :MFCD00063750
- :154.25 g/mol
- :286 °C (1013 hPa)
- :82…83 °C
- :Kamertemperatuur
- :229-998-8
- Verpakking:Glass bottle for solids
- VPE:1 g