Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Columns, MXT®-5

Columns, MXT®-5

4,5" standard coil, Capillair, MXT®-5, 0,20 µm, 0,18 mm, 40 m
Catalog # REST71823
Supplier:   Restek
Columns, MXT®-5
Catalog # REST71823
Supplier:  Restek
Columns, MXT®-5
Columns, MXT®-5

4,5" standard coil, Capillair, MXT®-5, 0,20 µm, 0,18 mm, 40 m

Price
1 460,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These MXT-5 columns are Siltek-treated stainless steel with low-polarity phase, crossbond diphenyl dimethyl polysiloxane stationary phase. All residual catalysts and low molecular weight fragments are removed from the MXT-5 polymer, providing a tight monomodal distribution and extremely low bleed.

  • Equivalent to USP G27, G36 phases
  • 4,5 inch standard coil diameter

General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, and semivolatiles.

Specifications
  • Kolomtype:
    Capillair
  • Filmdikte:
    0,20 µm
  • Ø inw.:
    0,18 mm
  • Omschrijving:
    4,5" standard coil
  • Lengte:
    40 m
  • Seq:
    05
  • Fase:
    MXT®-5