Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Columns, MXT®-5

Columns, MXT®-5

4,5" standard coil, Capillair, MXT®-5, 3,00 µm, 0,53 mm, 15 m
Catalogus # REST70282
Leverancier:   Restek
Columns, MXT®-5
Catalogus # REST70282
Leverancier:  Restek
Columns, MXT®-5
Columns, MXT®-5

4,5" standard coil, Capillair, MXT®-5, 3,00 µm, 0,53 mm, 15 m

Price
617,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

These MXT-5 columns are Siltek-treated stainless steel with low-polarity phase, crossbond diphenyl dimethyl polysiloxane stationary phase. All residual catalysts and low molecular weight fragments are removed from the MXT-5 polymer, providing a tight monomodal distribution and extremely low bleed.

  • Equivalent to USP G27, G36 phases
  • 4,5 inch standard coil diameter

General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, and semivolatiles.

Specificaties
  • Kolomtype:
    Capillair
  • Filmdikte:
    3,00 µm
  • Ø inw.:
    0,53 mm
  • Omschrijving:
    4,5" standard coil
  • Lengte:
    15 m
  • Seq:
    37
  • Fase:
    MXT®-5
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.