Columns, MXT®-5
4,5" standard coil, Capillair, MXT®-5, 5,00 µm, 0,53 mm, 30 m
Catalogus # REST70279
Leverancier: Restek
Ga naar:
Product Details & Documents
These MXT-5 columns are Siltek-treated stainless steel with low-polarity phase, crossbond diphenyl dimethyl polysiloxane stationary phase. All residual catalysts and low molecular weight fragments are removed from the MXT-5 polymer, providing a tight monomodal distribution and extremely low bleed.
- Equivalent to USP G27, G36 phases
- 4,5 inch standard coil diameter
General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, and semivolatiles.
Specificaties
- Kolomtype:Capillair
- Filmdikte:5,00 µm
- Ø inw.:0,53 mm
- Omschrijving:4,5" standard coil
- Lengte:30 m
- Seq:42
- Fase:MXT®-5
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



