Columns, MXT®-5
4,5" standard coil, Capillair, MXT®-5, 1,00 µm, 0,53 mm, 60 m
Catalog # REST70258
Supplier: Restek
Jump to:
Product Details & Documents
These MXT-5 columns are Siltek-treated stainless steel with low-polarity phase, crossbond diphenyl dimethyl polysiloxane stationary phase. All residual catalysts and low molecular weight fragments are removed from the MXT-5 polymer, providing a tight monomodal distribution and extremely low bleed.
- Equivalent to USP G27, G36 phases
- 4,5 inch standard coil diameter
General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, and semivolatiles.
Specifications
- Kolomtype:Capillair
- Filmdikte:1,00 µm
- Ø inw.:0,53 mm
- Omschrijving:4,5" standard coil
- Lengte:60 m
- Seq:33
- Fase:MXT®-5



