Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Columns, MXT®-5

4,5" standard coil, Capillair, MXT®-5, 0,50 µm, 0,25 mm, 30 m
  REST70238
 :   Restek
Columns, MXT®-5
  REST70238
 :  Restek
Columns, MXT®-5
Columns, MXT®-5

4,5" standard coil, Capillair, MXT®-5, 0,50 µm, 0,25 mm, 30 m

 
957,00 €
   
1 * 1 ST
 

These MXT-5 columns are Siltek-treated stainless steel with low-polarity phase, crossbond diphenyl dimethyl polysiloxane stationary phase. All residual catalysts and low molecular weight fragments are removed from the MXT-5 polymer, providing a tight monomodal distribution and extremely low bleed.

  • Equivalent to USP G27, G36 phases
  • 4,5 inch standard coil diameter

General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, and semivolatiles.

 
  • Kolomtype:
    Capillair
  • Filmdikte:
    0,50 µm
  • Ø inw.:
    0,25 mm
  • Omschrijving:
    4,5" standard coil
  • Lengte:
    30 m
  • Seq:
    20
  • Fase:
    MXT®-5