Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Columns, MXT®-5

4,5" standard coil, Capillair, MXT®-5, 0,25 µm, 0,25 mm, 15 m
Catalogus # REST70220
Leverancier:   Restek
Columns, MXT®-5
Catalogus # REST70220
Leverancier:  Restek
Columns, MXT®-5
Columns, MXT®-5

4,5" standard coil, Capillair, MXT®-5, 0,25 µm, 0,25 mm, 15 m

Price
591,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

These MXT-5 columns are Siltek-treated stainless steel with low-polarity phase, crossbond diphenyl dimethyl polysiloxane stationary phase. All residual catalysts and low molecular weight fragments are removed from the MXT-5 polymer, providing a tight monomodal distribution and extremely low bleed.

  • Equivalent to USP G27, G36 phases
  • 4,5 inch standard coil diameter

General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, and semivolatiles.

Specificaties
  • Kolomtype:
    Capillair
  • Filmdikte:
    0,25 µm
  • Ø inw.:
    0,25 mm
  • Omschrijving:
    4,5" standard coil
  • Lengte:
    15 m
  • Seq:
    06
  • Fase:
    MXT®-5