Orders LinkContactUsLinkComponent
AccuGENE® PBS Buffers
AccuGENE™ 1X PBS buffer, pH 7,4, Plastic bottle, 1 L
Catalogus # 733-1644
Leverancier:   LONZA
AccuGENE® PBS Buffers
Catalogus # 733-1644
Leverancier:  LONZA
CAS-nummer:  
AccuGENE® PBS Buffers
AccuGENE™ 1X PBS buffer, pH 7,4, Plastic bottle, 1 L
CAS-nummer:  
Price
49,00 €
Log in voor beschikbaarheid
1 * 1 L
Product Details & Documents
AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
  • Ready to use solutions
  • Reliable manufactured according to strict quality control standards to ensure lot to lot consistency
  • Guaranteed DNase-, RNase- and protease-free
Phosphate buffered saline, or PBS, is isotonic and non-toxic to most cells making it an excellent buffer for biological experiments. The osmolarity and ion concentrations of the solutions match those of the human body, so is often used to maintain cells for manipulation outside of their regular growth environment during experiments. PBS is also used in solutions that disengage attached and clumped cells.
Each reagent is prepared with 18 megOhm water, filtered using a 0,2 micron filter, and filled into sterile bottles.
For Research or Manufacturing Use Only: Not for use in in vitro Diagnostic Procedures, drug use, or for administration to humans or animals.
Specificaties
Specificaties
  • Verpakking:
    Plastic bottle
  • VPE:
    1 L
  • Omschrijving:
    AccuGENE™ 1X PBS buffer, pH 7,4