Test Tubes, with screw cap
100 mm, 16 mm, GL 18
Catalogus # HECH42782045
Leverancier: GLASWARENFABRIK KARL HECHT
Ga naar:
Product Details & Documents
AR®/soda glass, with PP screw cap.
- Heavy wall
- Round bottom
- With thread GL18
Specificaties
- Ø uitw.:16 mm
- Schroefdraad:GL 18
- Lengte:100 mm
- Seq:01