Reference electrodes
Referentie-elektrode, weinig onderhoud, Ag/AgCl-referentiesysteem, B 1181 HD, −5...+50 °C, Glas, Pasta, Keramisch, 12×120 mm, Schroefplugkop
Catalogus # SCOI285111701
Leverancier: SI Analytics
Ga naar:
Product Details & Documents
These reference electrodes are available with low maintenance plug heads or with fixed cables and free end.
Documents
SI Analytics Process Equipment Catalog VWR Process Equipment Catalog.pdf
Product documentatie
Specificaties
- Aansluiting:Schroefplugkop
- Ø×L:12×120 mm
- Type:B 1181 HD
- Omschrijving:Referentie-elektrode, weinig onderhoud, Ag/AgCl-referentiesysteem
- Materiaal:Glas
- Diafragma:Keramisch
- Elektroliet:Pasta
- Temperatuur:−5...+50 °C