Microplates, opaque OptiPlates™
OptiiPlate™-96 microplates, 96, wit, Case of 50
Catalogus # PERK6005290
Marketsource
Leverancier: PerkinElmer
Ga naar:
Product Details & Documents
These plates are used for general isotopic, fluorescence, LANCE™, [FP]², AlphaScreen™ or luminescence assays.
- Made of polystyrene
Specificaties
- Aant. wells:96
- Verpakt:Case of 50
- Kleur:wit
- Omschrijving:OptiiPlate™-96 microplates
- Seq:02