PH-combinatie-elektroden
Sensolyt® DWA pH single rod measuring chain**, Sensolyt® DWA, for installation into the SensoLyt® fitting, 0 - 14, Drinking water
Catalogus # 708-0058
Leverancier: WTW ONLINE
Ga naar:
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
Specificaties
- Voor:Drinking water
- Type:Sensolyt® DWA, for installation into the SensoLyt® fitting
- Omschrijving:Sensolyt® DWA pH single rod measuring chain**
- Seq:37
- pH:0 - 14