Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
PH-combinatie-elektroden
PH-combinatie-elektroden
Sensolyt® DWA pH single rod measuring chain**, Sensolyt® DWA, for installation into the SensoLyt® fitting, 0 - 14, Drinking water
Catalogus # 708-0058
Leverancier:   WTW ONLINE
PH-combinatie-elektroden
Catalogus # 708-0058
Leverancier:  WTW ONLINE
PH-combinatie-elektroden
PH-combinatie-elektroden
Sensolyt® DWA pH single rod measuring chain**, Sensolyt® DWA, for installation into the SensoLyt® fitting, 0 - 14, Drinking water
Price
310,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
  • Ideal for measurements in water and drinking water
  • Also for high pressure and temperature conditions
  • Easy to maintain by easy measuring chain replacement
Specificaties
  • Voor:
    Drinking water
  • Type:
    Sensolyt® DWA, for installation into the SensoLyt® fitting
  • Omschrijving:
    Sensolyt® DWA pH single rod measuring chain**
  • Seq:
    37
  • pH:
    0 - 14
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.