Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Natriumhydroxide 8% in aqueous solution, extra pure
Plastic bottle, 10 L
  KRAF05197.6020
Natriumhydroxide 8% in aqueous solution, extra pure
  KRAF05197.6020
 :  1310-73-2
Natriumhydroxide 8% in aqueous solution, extra pure
Plastic bottle, 10 L
 :  1310-73-2
212,00 €
1 * 10 L
 :  Caustic soda
  •  :1310-73-2
  •  :NaOH
  •  :MFCD00003548
  •  :40 g/mol
  •  :119…135 °C (1013 hPa)
  •  :9…10 °C
  •  :1,53 g/cm³ (20 °C)
  •  :Kamertemperatuur
  •  :215-185-5
  •  :1824
  •  :8,II
  • Verpakking:
    Plastic bottle
  • VPE:
    10 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.