Orders LinkContactUsLinkComponent
Drip closures
Drip closures
Vertical dropper for aqueous solutions; White tamper evident closure with dropper insert for openings according to DIN-18 for low surface tension aqueous solutions; also containing ethanol up to 15%
Catalogus # 215-3814
Leverancier:   MUELLER KREMPEL
Drip closures
Catalogus # 215-3814
Leverancier:  MUELLER KREMPEL
Drip closures
Drip closures
Vertical dropper for aqueous solutions; White tamper evident closure with dropper insert for openings according to DIN-18 for low surface tension aqueous solutions; also containing ethanol up to 15%
Price
33,50 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents
Specificaties
  • Omschrijving:
    Vertical dropper for aqueous solutions; White tamper evident closure with dropper insert for openings according to DIN-18 for low surface tension aqueous solutions; also containing ethanol up to 15%