Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
pH bufferoplossingen, Pure Water™
pH bufferoplossingen, Pure Water™
pH buffer A, (475 ml bottle), 6,97, Plastic bottle, 4
  662-0976
pH bufferoplossingen, Pure Water™
  662-0976
 :  
pH bufferoplossingen, Pure Water™
pH bufferoplossingen, Pure Water™
pH buffer A, (475 ml bottle), 6,97, Plastic bottle, 4
 :  
385,00 €
1 * 4 ST
Buffers for calibration giving fast, accurate pH results in high purity and low ionic strength waters. Recommended for samples such as well water, rain water, distilled/deionized water, boiler feed water, and process water.
  • Increases measurement accuracy
  • Reduces electrode drift
  • Eliminates contamination of sample
  • For best results, a high quality pH electrode is recommended
pHISA™Adjustor additive is designed to increase sample ionic strength without changing the pH of the sample.
  • Verpakking:
    Plastic bottle
  • VPE:
    4
  • Omschrijving:
    pH buffer A, (475 ml bottle)
  • pH-waarde:
    6,97