pH bufferoplossingen, Pure Water™
pH buffer A, (475 ml bottle), 6,97, Plastic bottle, 4
662-0976
Buffers for calibration giving fast, accurate pH results in high purity and low ionic strength waters. Recommended for samples such as well water, rain water, distilled/deionized water, boiler feed water, and process water.
- Increases measurement accuracy
- Reduces electrode drift
- Eliminates contamination of sample
- For best results, a high quality pH electrode is recommended
pHISA™Adjustor additive is designed to increase sample ionic strength without changing the pH of the sample.
- Verpakking:Plastic bottle
- VPE:4
- Omschrijving:pH buffer A, (475 ml bottle)
- pH-waarde:6,97