Celtellers, Sedgewick Rafter
Celteller, precisietype, glas met chroomafzetting, geslepen en afgeschuinde randen, Sedgewick Rafter, 1 mm, 50×20 mm
Catalog # MARI0611300
Supplier: MARIENFELD
Jump to:
Product Details & Documents
Counting chambers according to Sedgewick for particles and microorganisms in transparent liquids.
A cell is ruled in a grid subdividing 1,0 ml into 1000 μl.
Delivery information: Supplied with a cover glass.
Specifications
- Kamerdiepte:1 mm
- B×D:50×20 mm
- Omschrijving:Celteller, precisietype, glas met chroomafzetting, geslepen en afgeschuinde randen, Sedgewick Rafter
Accessories
Product Information
- Catalog #OmschrijvingAvailabilityPrice