Weegschuitjes
6 ml, 95 mm
Catalogus # 611-9310
Leverancier: Lenz Laborglas
Ga naar:
Product Details & Documents
Borosilicaatglas.
- Ideal for weighing small samples
- The tubular stem of the scoop also works as a weighing funnel and facilitates emptying of the scoop
Documents
BSE TSE Glass Columns
BSE/TSE-certificaten
Specificaties
- Capaciteit:6 ml
- Lengte:95 mm