Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
NS-stoppen
NS-stoppen
45/40, Flat
Catalogus # LENZ01400345
Leverancier:   Lenz Laborglas
NS-stoppen
Catalogus # LENZ01400345
Leverancier:  Lenz Laborglas
NS-stoppen
NS-stoppen
45/40, Flat
Price
112,00 €
Log in voor beschikbaarheid
1 * 5 ST
Product Details & Documents
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
  • Excellent chemical resistance
  • High thermal resistance, minimal heat expansion results in high resistance to temperature changes
  • Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
Certificeringen: DIN 12252
Bestelinformatie: Available with flat or pointed bottom.
Documents
Specificaties
  • (NS):
    45/40
  • Type:
    Flat
  • Seq:
    18
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.