Screw caps for screw thread tubes
GL 14
Catalogus # LENZ01320014
Leverancier: Lenz Laborglas
Ga naar:
Product Details & Documents
PBT,red, with PTFE coated silicone sealing (VMQ).
- For DIN GL threads
- Heat resistant up to 180 °C
- Autoclavable, sterilisable with hot air
Specificaties
- Schroefdraad:GL 14
- Seq:01