Orders LinkContactUsLinkComponent
Accessories for comparators
Colour (Gardner/APHA), 4/30BS, 10,0 - 18,0 pH
Catalogus # TINT243040
Leverancier:   Lovibond Tintometer
Accessories for comparators
Catalogus # TINT243040
Leverancier:  Lovibond Tintometer
Accessories for comparators
Accessories for comparators
Colour (Gardner/APHA), 4/30BS, 10,0 - 18,0 pH
Price
498,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+
Specificaties
  • Meetbereik:
    10,0 - 18,0 pH
  • Seq:
    82
  • Code:
    4/30BS
  • Testmethode:
    Colour (Gardner/APHA)
Recommendations