Accessories for comparators
Colour (Gardner/APHA), 4/30AS, 1,0 - 9,0 pH
Catalog # TINT243030
Supplier: Lovibond Tintometer
Jump to:
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
- Disc codes starting with 'N' are for use with the Nessleriser 2150
- Disc codes starting with 'C' are for use with the Nessleriser 2250
- All other discs are for use with the Comparator 2000+
Specifications
- Meetbereik:1,0 - 9,0 pH
- Seq:81
- Code:4/30AS
- Testmethode:Colour (Gardner/APHA)