Orders LinkContactUsLinkComponent
Accessories for comparators
Accessories for comparators
Colour (Gardner/APHA), 4/30AS, 1,0 - 9,0 pH
Catalog # TINT243030
Supplier:   Lovibond Tintometer
Accessories for comparators
Catalog # TINT243030
Accessories for comparators
Accessories for comparators
Colour (Gardner/APHA), 4/30AS, 1,0 - 9,0 pH
Price
498,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+
Specifications
  • Meetbereik:
    1,0 - 9,0 pH
  • Seq:
    81
  • Code:
    4/30AS
  • Testmethode:
    Colour (Gardner/APHA)