Orders LinkContactUsLinkComponent
Accessories for comparators
Accessories for comparators
AF332 EP colour scale, 4/64D, BY1 - BY7
Catalogus # TINT246440
Leverancier:   Lovibond Tintometer
Accessories for comparators
Catalogus # TINT246440
Leverancier:  Lovibond Tintometer
Accessories for comparators
Accessories for comparators
AF332 EP colour scale, 4/64D, BY1 - BY7
Price
427,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+
Specificaties
  • Meetbereik:
    BY1 - BY7
  • Seq:
    86
  • Code:
    4/64D
  • Testmethode:
    AF332 EP colour scale
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.