Cylinders, without lid
120 mm
Catalogus # HECH44409030
Leverancier: GLASWARENFABRIK KARL HECHT
Ga naar:
Product Details & Documents
Jars, made of glass or stainless steel, with base, without lids.
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Ordering information: Lids with knob available must be ordered separately.
Specificaties
- Ø uitw.:120 mm
- Seq:19