Dubbele spatels, breed
150 mm, 20 mm
Catalogus # RSGA092.150
Leverancier: RSG
Ga naar:
Product Details & Documents
Double spatulas with wide, flat endings. Made of stainless steel.
Documents
RSGA092.500_TDS_EN
Technische fiche
Specificaties
- Lengte:150 mm
- Breedte blad:20 mm