Orders LinkContactUsLinkComponent
Apothekerspatels
Apothekerspatels
Wooden handle, 130 mm, 20 mm
Catalog # 231-0320
Supplier:   RSG
Apothekerspatels
Catalog # 231-0320
Supplier:  RSG
Apothekerspatels
Apothekerspatels
Wooden handle, 130 mm, 20 mm
Price
132,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
Chromium steel, hardened, extremely flexible blade.
  • Excellent thermal resistance
  • Good chemical resistance
  • Quick and easy maintenance
  • Available with wooden handle or with dishwasher-safe plastic handle
Specifications
  • Omschrijving:
    Wooden handle
  • Breedte blad:
    20 mm
  • Lengte blad:
    130 mm