Apothekerspatels
Wooden handle, 150 mm, 22 mm
231-0321
: RSG
Chromium steel, hardened, extremely flexible blade.
- Excellent thermal resistance
- Good chemical resistance
- Quick and easy maintenance
- Available with wooden handle or with dishwasher-safe plastic handle
- Omschrijving:Wooden handle
- Breedte blad:22 mm
- Lengte blad:150 mm



