Dubbele spatels, micro, stijf
130 mm, 5 mm
Catalogus # RSGA835.130
Leverancier: RSG
Ga naar:
Product Details & Documents
Rigid micro double spatulas made of stainless steel.
Specificaties
- Lengte:130 mm
- Breedte blad:5 mm
Rigid micro double spatulas made of stainless steel.