Apothekerspatels
Wooden handle, 100 mm, 20 mm
Catalogus # 231-0319
Leverancier: RSG
Ga naar:
Product Details & Documents
Chromium steel, hardened, extremely flexible blade.
- Excellent thermal resistance
- Good chemical resistance
- Quick and easy maintenance
- Available with wooden handle or with dishwasher-safe plastic handle
Specificaties
- Omschrijving:Wooden handle
- Breedte blad:20 mm
- Lengte blad:100 mm



