Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Apothekerspatels

Wooden handle, 75 mm, 18 mm
Catalogus # 231-0318
Leverancier:   RSG
Apothekerspatels
Catalogus # 231-0318
Leverancier:  RSG
Apothekerspatels
Apothekerspatels

Wooden handle, 75 mm, 18 mm

Price
116,00 €
Log in voor beschikbaarheid
1 * 10 ST
Product Details & Documents

Chromium steel, hardened, extremely flexible blade.

  • Excellent thermal resistance
  • Good chemical resistance
  • Quick and easy maintenance
  • Available with wooden handle or with dishwasher-safe plastic handle
Specificaties
  • Omschrijving:
    Wooden handle
  • Breedte blad:
    18 mm
  • Lengte blad:
    75 mm