Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Apothekerspatels

Apothekerspatels

Plastic handle, dishwasher-safe, 130 mm, 20 mm
Catalogus # 231-0557
Leverancier:   RSG
Apothekerspatels
Catalogus # 231-0557
Leverancier:  RSG
Apothekerspatels
Apothekerspatels

Plastic handle, dishwasher-safe, 130 mm, 20 mm

Price
74,40 €
Log in voor beschikbaarheid
1 * 10 ST
Product Details & Documents

Chromium steel, hardened, extremely flexible blade.

  • Excellent thermal resistance
  • Good chemical resistance
  • Quick and easy maintenance
  • Available with wooden handle or with dishwasher-safe plastic handle
Specificaties
  • Omschrijving:
    Plastic handle, dishwasher-safe
  • Breedte blad:
    20 mm
  • Lengte blad:
    130 mm