Microtubes, with Thread
Micro tubes without screw cap, with skirted base, 2,0 ml, 5000/case
Catalog # SARS72.609
Marketsource
Supplier: SARSTEDT
Jump to:
Product Details & Documents
PP, translucent.
- Safe screw cap with O-ring
- Sterile or non sterile
- Autoclavable
For transport, storage and sample processing in the laboratory.
Specifications
- Verpakt:5000/case
- Capaciteit:2,0 ml
- Omschrijving:Micro tubes without screw cap, with skirted base
- Seq:22