Screw caps for micro tubes
Screw caps for micro tubes, wit
Catalogus # SARS65.716.022
Marketsource
Leverancier: SARSTEDT
Ga naar:
Product Details & Documents
PP, coloured.
- Excellent chemical resistance
- Excellent temperature resistance
Screw cap with O-ring seal.
Specificaties
- Kleur:wit
- Omschrijving:Screw caps for micro tubes
- Seq:11