BOD bottles
BOD bottles, with glass stopper, 300 ml
Catalogus # HACH62106
Leverancier: Hach
Ga naar:
Product Details & Documents
These are standard bottles, made of borosilicate glass, for BOD analaysis.
- Flared lip and ground-glass stopper to prevent air entrapment and for water seal
- Serialised bottles have silk-screened, sequential numbers and include a bar code
- Frosted marking area
Levering: Stoppers are included.
Specificaties
- Capaciteit:300 ml
- Omschrijving:BOD bottles, with glass stopper
- Seq:02