Orders LinkContactUsLinkComponent
SPE columns, CHROMABOND® C8
SPE columns, CHROMABOND® C8
C8, 60 Å, 45 µm, 96×100 mg
Catalogus # MANA738021.100M
Leverancier:   MACHEREY-NAGEL
SPE columns, CHROMABOND® C8
Catalogus # MANA738021.100M
Leverancier:  MACHEREY-NAGEL
SPE columns, CHROMABOND® C8
SPE columns, CHROMABOND® C8
C8, 60 Å, 45 µm, 96×100 mg
Price
291,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
CHROMABOND® C8 is an octyl modified silica phase for SPE.
  • Slightly more polar than C18 phase
  • Hydrophobic and more pronounced secondary interactions with polar compounds
  • Suitable for slight nonpolar and polar substances
Recommended for polar pesticides and PCBs.
Specificaties
  • Partikelgrootte:
    45 µm
  • Poriëngrootte:
    60 Å
  • Seq:
    09
  • Bedgewicht:
    96×100 mg
  • Fase:
    C8