Orders LinkContactUsLinkComponent
Empty solid phase extraction columns, CHROMABOND®
Empty PP columns with PE frits, one filter element is already inserted in the PP column, 150 ml
  MANA730474
Empty solid phase extraction columns, CHROMABOND®
  MANA730474
Empty solid phase extraction columns, CHROMABOND®
Empty solid phase extraction columns, CHROMABOND®
Empty PP columns with PE frits, one filter element is already inserted in the PP column, 150 ml
172,00 €
1 * 20 ST
Empty and reservoir columns with accessories for solid phase extraction.
  • Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
  • Reservoir columns from PP for application of medium-size samples
  • Accessories for SPE columns (filter elements, adapters)
  • Capaciteit:
    150 ml
  • Omschrijving:
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
  • Seq:
    08
Accessories
  •  
    Omschrijving
  • MANA732019
    PE frits for LV PP columns 15 ml, for 100 mg adsorbent weight
    1 * 250 ST - 132,00 €
  •  
    Omschrijving
  • MANA732020
    PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weight
    1 * 250 ST - 185,00 €
  •  
    Omschrijving
  • MANA730101
    Adapters (PP) for PP columns (1, 3 and 6 ml)
    1 * 10 ST - 96,10 €
  •  
    Omschrijving
  • MANA730105
    Adapters (PVDF) for glass columns (3 and 6 ml)
    1 * 10 ST - 200,00 €
  •  
    Omschrijving
  • MANA730026
    PE frits for PP columns, 70 ml
    1 * 250 ST - 281,00 €
  •  
    Omschrijving
  • MANA730192
    Glass fiber frits for glass columns, 6 ml
    1 * 250 ST - 238,00 €
  •  
    Omschrijving
  • MANA730351
    PE frits for PP columns, 15 ml
    1 * 250 ST - 238,00 €
  •  
    Omschrijving
  • MANA730356
    PE frits for PP columns, 45 ml
    1 * 250 ST - 273,00 €
  •  
    Omschrijving
  • MANA730475
    PE frits for PP columns, 150 ml
    1 * 250 ST - 302,00 €
  •  
    Omschrijving
  • MANA730034
    PE frits for PP columns, 30 ml
    1 * 250 ST - 258,00 €
  •  
    Omschrijving
  • MANA730566
    Adapters (PE) for PP columns (30 and 70 ml)
    1 * 1 KIT - 29,10 €
  •  
    Omschrijving
  • MANA730385
    Adapters (PE) for PP columns (15, 45 and 70 ml)
    1 * 10 ST - 125,00 €
  •  
    Omschrijving
  • MANA730191
    Glass fiber frits for glass columns, 3 ml
    1 * 250 ST - 196,00 €
  •  
    Omschrijving
  • MANA730164
    PE frits for PP columns, 1 ml
    1 * 250 ST - 130,00 €
  •  
    Omschrijving
  • MANA730163
    PE frits for PP columns, 6 ml
    1 * 250 ST - 226,00 €
  •  
    Omschrijving
  • MANA730162
    PE frits for PP columns, 3 ml
    1 * 250 ST - 184,00 €