Orders LinkContactUsLinkComponent
ATC probes, Eutech
ATC probes, Eutech
ATC probe, 1 m cable, pH 5+ / pH 6+ / ION 6+ meters
Catalogus # EUTEECPH5TEM01P
Leverancier:   EUTECH
ATC probes, Eutech
Catalogus # EUTEECPH5TEM01P
Leverancier:  EUTECH
ATC probes, Eutech
ATC probes, Eutech
ATC probe, 1 m cable, pH 5+ / pH 6+ / ION 6+ meters
Price
72,05 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
ATC electrodes for use with Eutech pH 5+, pH 6+ and ION 6+ meters or for use with CyberScan pH 300 and 310 meters.
Specificaties
  • Voor:
    pH 5+ / pH 6+ / ION 6+ meters
  • Omschrijving:
    ATC probe, 1 m cable