Orders LinkContactUsLinkComponent
Stoppers, flat type, PTFE
19/38, 50 mm
Catalogus # COWI010.119
Leverancier:   COWIE
Stoppers, flat type, PTFE
Catalogus # COWI010.119
Leverancier:  COWIE
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
19/38, 50 mm
Price
17,90 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
PTFE. Stopper with flat head fit standard ground glass joints.
  • Trapezoidal rings give high vacuum performance and reduce seizure
Specificaties
  • (NS):
    19/38
  • Hoogte:
    50 mm