Orders LinkContactUsLinkComponent
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
29/42, 57 mm
Catalogus # COWI010.129
Leverancier:   COWIE
Stoppers, flat type, PTFE
Catalogus # COWI010.129
Leverancier:  COWIE
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
29/42, 57 mm
Price
23,90 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
PTFE. Stopper with flat head fit standard ground glass joints.
  • Trapezoidal rings give high vacuum performance and reduce seizure
Specificaties
  • (NS):
    29/42
  • Hoogte:
    57 mm