Orders LinkContactUsLinkComponent
Jars with screw cap
Jars with screw cap
with polypropylene screw cap, individually wrapped, labelled, 150 ml
Catalogus # 216-2051
Leverancier:   Anicrin
Jars with screw cap
Catalogus # 216-2051
Leverancier:  Anicrin
Jars with screw cap
Jars with screw cap
with polypropylene screw cap, individually wrapped, labelled, 150 ml
Price
94,30 €
Log in voor beschikbaarheid
1 * 250 ST
Product Details & Documents
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
  • Twist cap for quick opening and closing
  • Very good temperature resistance
  • Very good chemical resistance
Let op: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Specificaties
  • Type:
    with polypropylene screw cap, individually wrapped, labelled
  • Capaciteit:
    150 ml
  • Seq:
    07