Handheld refractometers
Handheld refractometer, HSR-500, 0,0 − 90,0 °Bx, 0,2 °Brix
Catalogus # ATAG2340
Leverancier: Atago
Ga naar:
Product Details & Documents
These handheld refractometers provide three stages of scale - Calibrated with water.
: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Specificaties
- Model:HSR-500
- Meetbereik:0,0 − 90,0 °Bx
- Resolutie:0,2 °Brix
- Omschrijving:Handheld refractometer
- Cat. No.:ATAG2340
- b×d×h:40×40×20 mm
- Gewicht:600 g