Abbe refractometers, NAR
NAR-4T, 0 − 95 °Bx, 1,47 − 1,87 nD
Catalog # ATAG1240
Supplier: Atago
Jump to:
Product Details & Documents
The NAR Abbe refractometers can meet many of the requirements for routine measurements.
Single wavelengths and standard temperatures can be measured at extended ranges, as well as elevated temperatures, measurements of solid samples (films, plastic sheets).
Services: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Specifications
- Model:NAR-4T
- Meetbereik:0 − 95 °Bx
- Brekingsindex:1,47 − 1,87 nD
- Cat. No.:ATAG1240
- Temperatuurmeting:5...50 °C
- Temperatuursensor:±0,2 °C
- Nauwkeurigheid temperatuursensor:±0,2 °C
- Lichtbron:LED's
Accessories
Product Information
- Catalog #OmschrijvingAvailabilityPrice
Product Information
- Catalog #OmschrijvingAvailabilityPrice
- ATAGRE-1165Thermoweergave-eenheid (DTM-1), voor NAR-1T•LO, NAR-2T•LO en NAR-2T•HILog in for availability1 * 1 ST
Product Information
- Catalog #OmschrijvingAvailabilityPrice
Product Information
- Catalog #OmschrijvingAvailabilityPrice