Orders LinkContactUsLinkComponent
VWR®, Yeast extract
VWR®, Yeast extract
Yeast extract, 100 g
Catalogus # J850-100G
Leverancier:   VWR Chemicals
VWR®, Yeast extract
Catalogus # J850-100G
Leverancier:  VWR Chemicals
CAS-nummer:  
VWR®, Yeast extract
VWR®, Yeast extract
Yeast extract, 100 g
CAS-nummer:  
Price
111,00 €
Log in voor beschikbaarheid
1 * 100 g
Product Details & Documents
Yeast extract is the water soluble portion of autolysed yeast containing a source of Vitamin B complex.
  • High purity extract
  • Used for supplementing media
Specificaties
Specificaties
  • VPE:
    100 g
  • Omschrijving:
    Yeast extract
Specificaties