Orders LinkContactUsLinkComponent
VWR®NucleasEliminator™, Decontamination solution
VWR®NucleasEliminator™, Decontamination solution
NucleasEliminator™, impregnated wipes, 25
Catalogus # E891-25PK
Leverancier:   VWR Chemicals
VWR®NucleasEliminator™, Decontamination solution
Catalogus # E891-25PK
Leverancier:  VWR Chemicals
CAS-nummer:  
VWR®NucleasEliminator™, Decontamination solution
VWR®NucleasEliminator™, Decontamination solution
NucleasEliminator™, impregnated wipes, 25
CAS-nummer:  
Price
102,00 €
Log in voor beschikbaarheid
1 * 25 ST
Product Details & Documents
DNase and RNase contamination ruins expensive samples and time-consuming protocols in research laboratories every day. NucleasEliminator™ can help protect your investment by removing and deactivating even high concentrations of nuclease contamination from glass or plastic surfaces.
  • Effectively removes and deactivates nuclease contamination, even at high concentrations
  • Safe on all laboratory glass and plastic ware
  • Stable at room temperature for at least two years
Simply wipe, immerse, or fill the surface or vessel to be decontaminated with NucleasEliminator™, incubate 5 minutes at room temperature, and rinse thoroughly with nuclease-free water. NucleasEliminator™ is safe to use on all surfaces at the laboratory bench ranging from glassware, microcentrifuge tubes, pipette tips, and electrophoresis units.
A 100 ml bottle of the ready to use liquid will treat up to 100 microcentrifuge tubes or 25 casting trays for agarose electrophoresis.
Specificaties
Specificaties
  • VPE:
    25
  • Omschrijving:
    NucleasEliminator™, impregnated wipes
Specificaties