Tween® 80, Proteomics Grade
Cube container, 4 L
Fournisseur: VWR Chemicals
Jump to:
Product Details & Documents
Non-ionic detergent efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.
- CAS Number:9005-65-6
- MDL Number:MFCD00082107
- Merck Index:13,07664
- Boiling Pt:>100 °C (1013 hPa)
- Melting Pt:–21 °C
- Density:1,062 g/cm³ (20 °C)
- Flash Pt:149 °C (gesloten beker)
- Storage Temperature:Kamertemperatuur
- EINECS:500-019-9
Documents
Specifications
Specification Test Results
- DNase:None
- Heavy metals:<0.001%
- Hydroxyl number:65 − 80
- Identification:Pass
- Moisture (KF):3.0%
- Protease:None
- Residue on ignition:0.25%
- RNase:None
- Specific gravity:1.06 − 1.09
- Viscosity:300 − 500 centistokes
Specifications
- Verpakking:Cube container
- VPE:4 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



