Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Natriumhydroxide, beads, Reagent Grade

Natriumhydroxide, beads, Reagent Grade

HDPE bottle, 5 kg
Catalogus # 0173-5KG
Corrosief
Leverancier:   VWR Chemicals
Natriumhydroxide, beads, Reagent Grade
Catalogus # 0173-5KG
Leverancier:  VWR Chemicals
CAS-nummer:  1310-73-2
Natriumhydroxide, beads, Reagent Grade
Natriumhydroxide, beads, Reagent Grade

HDPE bottle, 5 kg

CAS-nummer:  1310-73-2
Price
506,00 €
Log in voor beschikbaarheid
1 * 5 kg
Product Details & Documents
Synoniemen:  Caustic soda
  • CAS-nummer:1310-73-2
  • Formule:NaOH
  • MDL-nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck index:14,08627
  • Kokend Pt:1390 °C (1013 hPa)
  • Smelten Pt:323 °C
  • Dichtheid:2,13 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • BEREIKEN:01-2119457892-27
Specificaties

Specificatie testresultaten

  • Chloride:
    ≤0.05%
  • Heavy Metals:
    ≤0.006%
  • Potassium:
    ≤0.1%
  • Purity:
    >98.5%
  • Sodium Carbonate:
    ≤0.90%
  • Sulphate:
    ≤0.005%

Specificaties

  • Verpakking:
    HDPE bottle
  • VPE:
    5 kg

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.