Stoppers, rubber, red, different sizes
25,0 mm, 17,0 mm, 22,0 mm
Catalogus # 146-0300
Leverancier: DEUTSCH NEUMANN
Ga naar:
Product Details & Documents
Specificaties
- Ø onder:17,0 mm
- Ø boven:22,0 mm
- Hoogte:25,0 mm
- Seq:06