Druppelflessen, WHEATON®
15-415, rood
WHEAW242512-A
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
- Schroefdraad:15-415
- Seq:33
- Kleur dop:rood
Screw cap made of radiation grade polypropylene.