Orders LinkContactUsLinkComponent
Druppelflessen, WHEATON®
Druppelflessen, WHEATON®
8 - 425, oranje
Catalogus # 215-4038
Leverancier:   DWK Life Sciences
Druppelflessen, WHEATON®
Catalogus # 215-4038
Leverancier:  DWK Life Sciences
Druppelflessen, WHEATON®
Druppelflessen, WHEATON®
8 - 425, oranje
Price
370,00 €
Log in voor beschikbaarheid
1 * 1.000 ST
Product Details & Documents
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
Specificaties
  • Schroefdraad:
    8 - 425
  • Seq:
    11
  • Kleur dop:
    oranje