BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 49-72, 300 ml
Catalogus # WHEA227497-03
Leverancier: DWK Life Sciences
Ga naar:
Product Details & Documents
Made of Type I borosilicate glass.
- Available with or without stopper
- With white labeling area
Specificaties
- Capaciteit:300 ml
- Omschrijving:BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 49-72