BOD bottles, WHEATON®
BOD bottles, borosilicate glass, 43×115 mm (Ø×H), number sequence: 73-108, 60 ml
Catalogus # WHEA227494-03
Leverancier: DWK Life Sciences
Ga naar:
Product Details & Documents
Made of Type I borosilicate glass.
- Available with or without stopper
- With white labeling area
Specificaties
- Capaciteit:60 ml
- Omschrijving:BOD bottles, borosilicate glass, 43×115 mm (Ø×H), number sequence: 73-108